Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A-RAF Antibody (M289), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323355
Description
A-RAF Monoclonal antibody specifically detects A-RAF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
A-RAF | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
M289 | |
Western Blot 1:1000 | |
ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1 | |
Clone (M289) was generated from a sequence corresponding to amino acids in the N-terminal region of human A-Raf. This A-Raf sequence has high homology to rat and mouse A-Raf. [UniProt# P10398] | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
369 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction