Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A-Raf Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584719 Supplier Invitrogen™ Supplier No. PA584719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSTNRQQFYH SVQDLSGGSR QHEAPSNRPL NELLTPQGPS PRTQHCDPEH FPFPAPANAP LQRIRSTST Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 81%.

Several serine/threonine protein kinases have been implicated as intermediates in signal transduction pathways. These include ERK/MAP kinases, ribosomal S6 kinase (Rsk) and Raf-1. Raf-1 is a cytoplasmic protein with intrinsic serine/threonine activity. It is broadly expressed in nearly all cell lines tested to date and is the cellular homolog of v-Raf, the product of the transforming gene of the 3611 strain of murine sarcoma virus. The unregulated kinase activity of the v-Raf protein has been associated with transformation and mitogenesis while the activity of Raf-1 is normally suppressed by a regulatory N-terminal domain. A-Raf, a second member of the Raf gene family of serine/ threonine protein kinases, exhibits substantial homology to Raf-1 within the kinase domain of the two molecules, but less homology elsewhere. Expression of A-Raf is found at highest levels in urogenital tissues and kidney and at lowest level in brain tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A-Raf
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARAF
Gene Accession No. P10398
Gene Alias 1200013E08Rik; Araf; A-RAF; A-Raf proto-oncogene serine/threonine-protein kinase; A-Raf proto-oncogene, serine/threonine kinase; Araf1; A-raf-1; AW495444; Oncogene ARAF1; PKS; PKS2; Proto-oncogene A-Raf; proto-oncogene A-Raf-1; Proto-oncogene Pks; raf a; Raf related protein; RAFA1; raf-related oncogene; Ras-binding protein; Ras-binding protein DA-Raf; RP1-230G1.1; serine/threonine-protein kinase A-Raf; v-raf murine sarcoma 3611 viral oncogene homolog; v-raf murine sarcoma 3611 viral oncogene homolog 1; v-raf murine sarcoma 3611 viral oncogene-like protein; v-raf oncogene homolog 1 (murine sarcoma 3611 virus)
Gene Symbols ARAF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARAF. Recombinant protein control fragment (Product #RP-103860).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 369
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.