Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ A-Raf Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587171
Description
Purity is > 95% by SDS-PAGE.
Several serine/threonine protein kinases have been implicated as intermediates in signal transduction pathways. These include ERK/MAP kinases, ribosomal S6 kinase (Rsk) and Raf-1. Raf-1 is a cytoplasmic protein with intrinsic serine/threonine activity. It is broadly expressed in nearly all cell lines tested to date and is the cellular homolog of v-Raf, the product of the transforming gene of the 3611 strain of murine sarcoma virus. The unregulated kinase activity of the v-Raf protein has been associated with transformation and mitogenesis while the activity of Raf-1 is normally suppressed by a regulatory N-terminal domain. A-Raf, a second member of the Raf gene family of serine/ threonine protein kinases, exhibits substantial homology to Raf-1 within the kinase domain of the two molecules, but less homology elsewhere. Expression of A-Raf is found at highest levels in urogenital tissues and kidney and at lowest level in brain tissue.
Specifications
A-Raf | |
Polyclonal | |
Unconjugated | |
ARAF | |
1200013E08Rik; Araf; A-RAF; A-Raf proto-oncogene serine/threonine-protein kinase; A-Raf proto-oncogene, serine/threonine kinase; Araf1; A-raf-1; AW495444; Oncogene ARAF1; PKS; PKS2; Proto-oncogene A-Raf; proto-oncogene A-Raf-1; Proto-oncogene Pks; raf a; Raf related protein; RAFA1; raf-related oncogene; Ras-binding protein; Ras-binding protein DA-Raf; RP1-230G1.1; serine/threonine-protein kinase A-Raf; v-raf murine sarcoma 3611 viral oncogene homolog; v-raf murine sarcoma 3611 viral oncogene homolog 1; v-raf murine sarcoma 3611 viral oncogene-like protein; v-raf oncogene homolog 1 (murine sarcoma 3611 virus) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11836, 369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P04627, P10398 | |
ARAF | |
Recombinant Protein of human ARAF. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction