Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A12B4C3
GREENER_CHOICESDP

Catalog No. 509710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-971-0 10 mg
50-975-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-971-0 Supplier Tocris Bioscience™ Supplier No. 5097/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor

A12B4C3 is a potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor (IC50 = 60 nM). Displays no effect on calcineurin, protein PP-1 or APTX. Interrupts DNA repair and enhances radiosensitivity of human A549 lung carcinoma and MDA-MB-231 cells.

Specifications

Quantity 10 mg
Product Type A12B4C3
Purity >98%
Molecular Weight (g/mol) 534.65
Inhibitors A12B4C3

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.