Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 4-ANPP Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Click to view available options
Quantity:
0.5 mL
Description
A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for 4-ANPP LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications. 4-ANPP is an intermediate in the synthesis of fentanyl and related opioids and is often found as an impurity in fentanyl preparations. This Certified Spiking Solution™ is supplied in a convenient US DEA-exempt solution format.

Specifications
Specifications
Packaging | Ampule |
CAS | 67-56-1 |
Flash Point | 9.7°C (49.5°F) - closed cup |
Linear Formula | C19H24N2 |
Concentration | 100 μg/mL in methanol |
MDL Number | MFCD00179023 |
UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
Quantity | 0.5 mL |
Formula Weight | 280.41 |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction