Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A1BG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588154 Supplier Invitrogen™ Supplier No. PA588154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSGVDFQLRR GEKELLVPRS STSPDRIFFH LNAVALGDGG HYTCRYRLHD NQNGWSGDSA PVELILSDET LPAPEFSPEP ESGRALRLRC LAPLEGARFA LVREDRGGRR VHRFQSPAGT EALFELHNIS VADSANYSCV YVDLKPPFGG SAPSERLELH VDGPPPRPQL RATWSGAVLA GRDAVLRCEG PIPDVTFELL REGETKAVKT VRTPGAAANL ELIFVGPQHA GNYRCRYRSW VPHTFESELS DPVELLVAES; Positive Samples: SKOV3, HeLa, Mouse kidney, Rat liver.

Alpha-1-B glycoprotein (A1BG) which is a plasma protein with no known function belongs to the immunoglobulin superfamily. Although the human A1BG has been known for four decades, and the information about the amino acid sequence, chromosomal assignment, and even genetic polymorphism in different populations have been known, no biological function has been suggested. A1BG (Mr approximately equal to 63,000) consists of one polypeptide chain of 474 amino acids with four glucosamine oligosaccharides. The polypeptide has five intrachain disulfide bonds and consists of five repeating structural domains, each containing about 95 amino acids and one disulfide bond. Recently human cysteine-rich secretory protein 3 (CRISP-3) has been found to be a binding partner of A1B, suggesting the A1BG-CRISP-3 complex displays a protecting function from a potentially harmful effect of free CRISP-3 in circulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A1BG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 11.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene A1BG
Gene Accession No. P04217
Gene Alias A1B; A1bg; A1bg {ECO:0000312; ABG; Alpha-1-B glycoprotein; alpha-1-B-glycoprotein; alpha-1B-glycoprotein; alpha1B-glycoprotein; C44; DKFZp686F0970; epididymis secretory sperm binding protein Li 163pA; GAB; HEL-S-163pA; HYST2477; Liver regeneration-related protein 1; RGD:69417}
Gene Symbols A1BG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 236-495 of human A1BG (NP_5706022).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.