Learn More
Invitrogen™ A1BG Polyclonal Antibody

Description
A1BG Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | A1BG |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | A1BG |
| Gene Accession No. | P04217 |
| Gene Alias | A1B; A1bg; A1bg {ECO:0000312; ABG; Alpha-1-B glycoprotein; alpha-1-B-glycoprotein; alpha-1B-glycoprotein; alpha1B-glycoprotein; C44; DKFZp686F0970; epididymis secretory sperm binding protein Li 163pA; GAB; HEL-S-163pA; HYST2477; Liver regeneration-related protein 1; RGD:69417} |
| Gene Symbols | A1BG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.