Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A1CF Polyclonal Antibody, Invitrogen™

Catalog No. PIPA541849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541849 Supplier Thermo Scientific Supplier No. PA541849

Rabbit Polyclonal Antibody

Peptide sequence: MESNHKSGDG LSGTQKEAAL RALVQRTGYS LVQENGQRKY GGPPPGWDAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%

A1CF Polyclonal antibody specifically detects A1CF in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A1CF
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene A1CF
Gene Accession No. Q9NQ94
Gene Alias 1810073H04Rik; A1cf; A1cft; Acf; ACF64; ACF65; apo-B RNA editing protein; Apobec-1; APOBEC1 complementation factor; apobec-1 complementation factor; apobec-1 complementation factor (ACF) (ASP); Apobec-1 complementation factor APOBEC-1 stimulating protein; Apobec-1 complementation factor, APOBEC-1 stimulating protein; APOBEC1 complementation factort; APOBEC-1 stimulating protein; APOBEC1CF; APOBEC1-stimulating protein; ASP; MGC163391; RP11-564C4.2
Gene Symbols A1CF
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human A1CF
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29974
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.