Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A20 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | A20 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-3077
![]() |
Bethyl Laboratories, Inc
A302633AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
A20/TNFAIP3 is an important regulator of proinflammatory signaling pathways, including NK-kB signaling. A20 is upregulated through NF-kB (p50/p55) activation, and subsequently down regulates NF-kB through its dual function as a deubiquitinase and ubiquitin ligase.Specifications
A20 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
A20, AISBL, OTU domain-containing protein 7C, OTUD7C, putative DNA-binding protein A20, TNF alpha-induced protein 3, TNFA1P2, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha induced protein 3, tumor necrosis factor, alpha-induced protein 3, zinc finger protein A20 | |
TNFAIP3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P21580 | |
7128 | |
Between 525 and 575 | |
Primary | |
4° C | |
TNFAIP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title