Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A20 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563075
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3075 Supplier Bethyl Laboratories, Inc Supplier No. A302633A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

A20/TNFAIP3 is an important regulator of proinflammatory signaling pathways, including NK-kB signaling. A20 is upregulated through NF-kB (p50/p55) activation, and subsequently down regulates NF-kB through its dual function as a deubiquitinase and ubiquitin ligase.

Specifications

Antigen A20
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TNFAIP3
Gene Accession No. P21580
Gene Alias A20, AISBL, OTU domain-containing protein 7C, OTUD7C, putative DNA-binding protein A20, TNF alpha-induced protein 3, TNFA1P2, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha induced protein 3, tumor necrosis factor, alpha-induced protein 3, zinc finger protein A20
Gene Symbols TNFAIP3
Host Species Rabbit
Immunogen Between 525 and 575
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7128
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.