Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A26C2/A26C3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA599692

Description
Antibody detects endogenous levels of total POTE14/22.
Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications
A26C2/A26C3 | |
Polyclonal | |
Unconjugated | |
POTEH | |
4921537P18Rik; 4930456F22Rik; A26C2; A26C3; ACTBL1; actin, beta-like 1; ANKRD26-like family C member 2; ANKRD26-like family C member 3; ANKRD26-like family C, member 2; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 4; cancer/testis antigen family 104, member 7; CT104.4; CT104.7; gonad specific ankyrin repeat protein; Gsarp1; Gsarp2; POTE ankyrin domain family member A; POTE ankyrin domain family member G; POTE ankyrin domain family member H; POTE ankyrin domain family, member G; POTE ankyrin domain family, member H; POTE14; POTE-14; POTE14alpha; POTE14B; POTE22; POTE-22; Poteg; POTEH; Prostate, ovary, testis-expressed protein on chromosome 14; prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 14; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like | |
Rabbit | |
Affinity chromatography | |
RUO | |
23784, 404785, 70952 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6S545, Q6S5H5 | |
POTEG, POTEH | |
A synthesized peptide derived from human POTE14/22, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction