Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2AR/Adenosine A2a receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983299
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3299 Supplier Bioss Supplier No. BS1456R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.

Specifications

Antigen A2AR/Adenosine A2a receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADORA2A
Gene Accession No. P29274
Gene Alias A2a receptor; A2AAR; A2aR; AA2AR; ADENO; adenosine A2a receptor; Adenosine A2a-receptor; Adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2; Adora2a; Adora2l1; AR; hA2aR; RDC8
Gene Symbols ADORA2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human A2a receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.