Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A2B5 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1416T100UG
This item is not returnable.
View return policy
Description
A2B5 Monoclonal specifically detects A2B5 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.Specifications
A2B5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat, Chicken | |
IgM |
Flow Cytometry | |
105 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Embryonic chicken retinal cells | |
RUO | |
Recognizes the A2B5 cell surface ganglioside epitope found on type II astrocytes, cells involved in gliogenesis, and cells committed to the oligodendrocyte lineage (1-4). | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction