Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A2B5 Monoclonal Antibody (105)
GREENER_CHOICE

Catalog No. PIMA190445
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190445 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190445 Supplier Invitrogen™ Supplier No. MA190445
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

A2B5 binds to neural tissue such as retina, brain, spinal cord, and dorsal root ganglia. It is also able to bind plasma membranes of human, rat, chicken and mouse islet cells and can be used in cell sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2B5
Applications Immunocytochemistry
Classification Monoclonal
Clone 105
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene Alias A2B5 marker
Host Species Mouse
Immunogen Embryonic chicken retinal cells.
Purification Method IgM
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.