Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A2BP1 Antibody (D8H8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP213169AF532
Description
A2BP1 Monoclonal specifically detects A2BP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| A2BP1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RBFOX1 | |
| Mouse recombinant A2BP1 [Swiss-ProtNo. Q9JJ43] | |
| 0.1 mL | |
| Primary | |
| Does not cross react with paralogue FOX2 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| D8H8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1 | |
| Mouse | |
| Protein G purified | |
| Epigenetics, Neuroscience | |
| 54715.0 | |
| Human, Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction