Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2LD1/GGACT Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981317
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1317 Supplier Bioss Supplier No. BS11381R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene aids in the proteolytic degradation of crosslinked fibrin by breaking down isodipeptide L-gamma-glutamyl-L-epsilon-lysine, a byproduct of fibrin degradation. The reaction catalyzed by the encoded gamma-glutamylaminecyclotransferase produces 5-oxo-L-proline and a free alkylamine. Two transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen A2LD1/GGACT
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GGACT
Gene Accession No. Q9BVM4
Gene Alias A2LD1; AIG2-like domain 1; AIG2-like domain-containing protein 1; BOS_12668; cDNA sequence BC006662-like; Gamma-glutamylamine cyclotransferase; Gamma-glutamylaminecyclotransferase; GGACT; LOC100355696; RGD1304748
Gene Symbols GGACT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human A2LD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 87769
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.