Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ A2M Monoclonal Antibody (A5-A9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA542577
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
Positive Control: LOVO cell lysates, LO2 cell lysates, Hela, HepG2, human liver tissue, human uterus tissue. Subcellular Location: Secreted. Product has been tested in direct ELISA.
Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregnancy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer's disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.
Specifications
A2M | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P01023 | |
A2M | |
Recombinant protein within human Alpha-2-macroglobulin aa 950-1200. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
A5-A9 | |
Unconjugated | |
A2M | |
A2M; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; LOC100061656; Mam; S863 7; S863-7 | |
Mouse | |
Protein G | |
RUO | |
2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction