Learn More
Invitrogen™ A2M Monoclonal Antibody (F1-P1C11 #3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MIA1201
Description
MIA1201 targets Alpha2-Macroglobulin in ELISA, immunoprecipitation, and radioimmunoassay applications and shows reactivity with Human samples. The MIA1201 immunogen is purified human plasma AMG. MIA1201 detects Alpha2-Macroglobulin which has a predicted molecular weight of approximately 162 kDa. MIA1201 was formerly sold as a Seradyn product.
Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregnancy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer's disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.
Specifications
A2M | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P01023 | |
A2M | |
Purified human plasma AMG. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
F1-P1C11 #3 | |
Unconjugated | |
A2M | |
A2M; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; LOC100061656; Mam; S863 7; S863-7 | |
Mouse | |
Protein A | |
RUO | |
2 | |
Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.