Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2M Mouse anti-Human, Clone: F1-P1C11 #3, Abnova™

Catalog No. 89035390 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-390 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-390 Supplier Abnova Corporation Supplier No. MAB13983

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human A2M.

  • High specificity
  • High reactivity

Specifications

Antigen A2M
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone F1-P1C11 #3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human A2M.
Dilution ELISA Immunoprecipitation Radioimmunoassay The optimal working dilution should be determined by the end user.
Formulation In PBS (1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P01023
Gene Alias CPAMD5/DKFZp779B086/FWP007/S863-7
Gene Symbols A2M
Host Species Mouse
Immunogen Native purified A2M from human plasma.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.