Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A2M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120391 Supplier Invitrogen™ Supplier No. PA5120391
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SH-SY5Y, Mouse kidney, Mouse ovary, Rat spinal cord. The target is usually found in the following locations: Secreted. Immunogen sequence: ALAGNQDKRK EVLKSLNEEA VKKDNSVHWE RPQKPKAPVG HFYEPQAPSA EVEMTSYVLL AYLTAQPAPT SEDLTSATNI VKWITKQQNA QGGFSSTQDT VVALHALSKY GAATFTRTGK AAQVTIQSSG TFSSKFQVDN NNRLLLQQVS LPELPGEYSM KVTGEGCVYL QTSLKYNILP EKEEFPFALG VQTLPQTCDE PKAHTSFQIS LSVSYTGSRS ASNMAIVDVK MVSGFIPLKP TVKMLERSNH VSRTEVSSNH VLIYLDKVSN QTLSLFFTVL QDVPVRDLKP AIVKVYDYYE TDEFAIAEYN APCSKDLGNA.

Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregnancy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer's disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2M
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene A2M
Gene Accession No. P01023, P06238, Q6GQT1
Gene Alias A2M; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; LOC100061656; Mam; S863 7; S863-7
Gene Symbols A2M
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2, 232345, 24153
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.