Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A2ML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558562 Supplier Invitrogen™ Supplier No. PA558562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTFEDTSNFY HPNFPFSGKI RVRGHDDSFL KNHLVFLVIY GTNGTFNQTL VTDNNGLAPF TLETSGWNGT DVSLEGKFQM EDLVYNPEQV PRYYQ Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 22%.

A2ML1 is a new member of the alpha 2-macroglobulin protease inhibitor family, and shares most the characteristics of this family. It is the first family member detected in the epidermis and may play an important role during desquamation by inhibiting extracellular proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2ML1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene A2ML1
Gene Accession No. A8K2U0
Gene Alias A2ML1; alpha-2-macroglobulin like 1; alpha-2-macroglobulin-like 1; alpha-2-macroglobulin-like protein 1; BC048546; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 9; C3 and PZP-like, alpha-2-macroglobulin domain containing 9; cDNA sequence BC048546; CPAMD9; Ovos; Ovostatin homolog
Gene Symbols A2ML1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human A2ML1. Recombinant protein control fragment (Product #RP-97812).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144568
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.