Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A33/GPA33 Antibody (022), Alexa Fluor™ 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289872AF405
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for ELISASpecifications
A33/GPA33 | |
Monoclonal | |
Alexa Fluor 405 | |
A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human A33/GPA33 (Q99795-1; Met1-Val235). | |
0.1 mL | |
Cancer | |
10223 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
022 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction