Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598802
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598802 Supplier Invitrogen™ Supplier No. PA598802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A33 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A33
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Gpa33
Gene Accession No. Q99795
Gene Alias 2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33
Gene Symbols Gpa33
Host Species Rabbit
Immunogen Recombinant Human Cell surface A33 antigen protein (22-235AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10223
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.