Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A37
GREENER_CHOICE

Catalog No. 580210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-021-0 10 mg
58-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-021-0 Supplier Tocris Bioscience™ Supplier No. 5802/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

ALDH1A1 inhibitor

ALDH1A1 inhibitor (IC50 = 4.6 μM; Ki = 300 nM). Exhibits selectivity for ALDH1A1 over eight other ALDH isoforms. Disrupts spheroid formation and reduces cell viability of ovarian cancer cells in vitro.

Specifications

Quantity 10 mg
Formula Weight 431.57
Physical Form Solid
Purity >97%
Chemical Name or Material Ethyl 2-[[3,4-dihydro-4-oxo-3-[3-(1-pyrrolidinyl)propyl][1]benzothieno[3,2-d]pyrimidin-2-yl]thio]ace
Recommended Storage Soluble to 100mM in DMSO and to 100mM in ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.