Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ KGA300 Coveralls with Hood and Boots
GREENER_CHOICE

Catalog No. 19005105 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-105 Medium
19-005-104 Small
19-005-106 Large
19-005-107 X-Large
19-005-108 2X-Large
19-005-109 3X-Large
19-005-110 4X-Large
19-005-111 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-005-105 Supplier Ansell Supplier No. 48972
Add to Cart
Add to Cart
This item is not returnable. View return policy

KGA300 coveralls for surface preparation and paint protection are designed to protect against hazardous particulates and certain light liquid splashes while providing a comfortable fit and freedom of movement.

  • KGA300 liquid and surface prep and paint protection coveralls help protect against hazardous particulates and certain light liquid splashes
  • Made of disposable, lightweight microporous film laminate
  • Pass NFPA 99 for antistatic materials
  • Feature serged seams and a water resistant front zipper, and come in a variety of sizes and configurations
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations
  • Zipper front, elastic wrists, ankles, attached hood and boots
  • Silicone- and latex-free
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Single-use
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Liquid, Particle
Coating Anti-static
Back Style Elastic
Certifications/Compliance NFPA 99, TAA Compliant Protective Apparel
Latex Free Yes
Model KleenGuard™ KGA300
For Use With (Application) Surface Preparation and Painting
Quantity 25/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.