Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A4GALT Monoclonal Antibody (38.13)
GREENER_CHOICE

Catalog No. PIMA544001
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544001 Supplier Invitrogen™ Supplier No. MA544001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

This antibody recognizes CD77 (globotriaosylceramide Gb3), a neutral glycosphingolipid expressed mainly on the surface of B cell populations, such as germinal center B cells and Burkitt's lymphoma cells.

The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A4GALT
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 38.13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 15mM sodium azide; pH 8
Gene A4galt
Gene Accession No. Q9NPC4
Gene Alias A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4Gal-T1; cd77; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase
Gene Symbols A4galt
Host Species Rat
Immunogen Daudi cell line (Burkitts lymphoma).
Purification Method Sequential chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53947
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.