Learn More
Invitrogen™ A4GALT Polyclonal Antibody

Description
The antibody detects endogenous levels of total CD77 / A14GALT protein.
Specifications
Specifications
| Antigen | A4GALT |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | A4galt |
| Gene Accession No. | Q9NPC4 |
| Gene Alias | A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4Gal-T1; cd77; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase |
| Gene Symbols | A4galt |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.