Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A4GALT Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987851
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7851 Supplier Bioss Supplier No. BS7588R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor.

Specifications

Antigen A4GALT
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene A4galt
Gene Accession No. Q9NPC4
Gene Alias A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; Alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; Alpha4Gal-T1; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; Lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase
Gene Symbols A4galt
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human A4GALT/CD77.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53947
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.