Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A4GNT Antibody (OTI2H9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271905AF700
Description
A4GNT Monoclonal specifically detects A4GNT in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
A4GNT | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
A4GNT | |
Recombinant protein fragment corresponding to amino acids 121-340 of human A4GNT (NP_057245) produced in E.coli. | |
0.1 mL | |
51146.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2H9 | |
Western Blot, Immunohistochemistry | |
alpha-1,4-N-acetylglucosaminyltransferase | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction