Learn More
A4GNT Mouse anti-Human, Clone: OTI5B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800400

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.Specifications
A4GNT | |
Monoclonal | |
Unconjugated | |
A4GNT | |
alpha4GnT | |
Mouse | |
Affinity Chromatography | |
RUO | |
51146 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
OTI5B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UNA3 | |
A4GNT | |
Human recombit protein fragment corresponding to amino acids 121-340 of human A4GNT produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.