Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ A4GNT Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519221
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
Specifications
| A4GNT | |
| Polyclonal | |
| Unconjugated | |
| A4GNT | |
| A4GNT; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; AV080780; EGK_12072; Gm798; GnTalpha4; MGC149493 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51146 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UNA3 | |
| A4GNT | |
| Synthetic peptide sequence (PRTYRDLIKGPEGS) corresponding to the C-terminus amino acids of A4GNT (aa 317-331). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction