Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A4GNT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588499 Supplier Invitrogen™ Supplier No. PA588499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRKELQLSLS VTLLLVCGFL YQFTLKSSCL FCLPSFKSHQ GLEALLSHRR GIVFLETSER MEPPHLVSCS VESAAKIYPE WPVVFFMKGL TDSTPMPSNS TYPAFSFLSA IDNVFLFPLD MKRLLEDTPL FSWYNQINAS AERNWLHISS DASRLAIIWK YGGIYMDTDV ISIRPIPEEN FLAAQASRYS SNGIFGFLPH HPFLWECMEN FVEHYNSAIW GNQGPELMTR MLRVWCKLED FQEVSDLRCL NISFLHPQRF YPISYREWRR YYEVWDTEPS FNVSYALHLW NHMNQEGRAV IRGSNTLVEN LYRKHCPRTY RDLIKGPEGS VTGELGPGNK; Positive Samples: SGC-7901, 293T, LO2; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A4GNT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene A4GNT
Gene Accession No. Q9UNA3
Gene Alias A4GNT; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; AV080780; EGK_12072; Gm798; GnTalpha4; MGC149493
Gene Symbols A4GNT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human A4GNT (NP_0572451).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51146
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.