Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AA 147
GREENER_CHOICESDP

Catalog No. 67595 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-595 5 mg
67-592-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-595 Supplier Tocris Bioscience™ Supplier No. 6759/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ER proteostasis regulator

AA 147 is an endoplasmic reticulum (ER) proteostasis regulator. Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation. Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells. Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).

Specifications

Chemical Name or Material N-(2-Hydroxy-5-methylphenyl)benzenepropanamide
CAS 393121-74-9
Quantity 5 mg
Molecular Formula C16H17NO2
Formula Weight 255.31
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.