Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    AA5 Virus VP3 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
AA5 Virus VP3 Polyclonal Antibody
Supplier: Thermo Scientific PA14108
Description
AA5 Virus VP3 Polyclonal Antibody for Western Blot
            Specifications
| AA5 Virus VP3 | |
| Polyclonal | |
| Unconjugated | |
| AAV; AAV-5 VP3; AdV5; AV5; HADV-5; Lentivirus | |
| Synthetic peptide corresponding to residues N(530) S Q P A N P G T T A T C(541) of the 80 kDa capsid VP3 protein of AAV 5. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid | 
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction