Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AA5 Virus VP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA123359
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123359 Supplier Invitrogen™ Supplier No. PA123359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-23359 detects adeno-associated 5 virus in virus samples.

Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AA5 Virus VP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias AAV; AAV-5 VP3; AdV5; AV5; HADV-5; Lentivirus
Host Species Rabbit
Immunogen Residues N(530) S Q P A N P G T T A T C(541) of the 80 kDa capsid VP3 protein of AAV5.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.