Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AAAS Antibody (OTI5B10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271358AF405

Catalog No. N271358A405


Only null left
Add to Cart

Description

Description

AAAS Monoclonal specifically detects AAAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

AAAS
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
AAAS
Recombinant protein fragment corresponding to amino acids 322-546 of human AAAS (NP_056480) produced in E.coli.
0.1 mL
8086.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI5B10
Western Blot, Immunohistochemistry
AAA, AAASb, achalasia, adrenocortical insufficiency, alacrimia, achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A), ADRACALA, Adracalin, ALADIN, Allgrove, triple-A, DKFZp586G1624
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.