Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAAS Antibody (OTI5B10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271358G
Description
AAAS Monoclonal specifically detects AAAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AAAS | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AAAS | |
Recombinant protein fragment corresponding to amino acids 322-546 of human AAAS (NP_056480) produced in E.coli. | |
0.1 mL | |
8086.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5B10 | |
Western Blot, Immunohistochemistry | |
AAA, AAASb, achalasia, adrenocortical insufficiency, alacrimia, achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A), ADRACALA, Adracalin, ALADIN, Allgrove, triple-A, DKFZp586G1624 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction