Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AACS Monoclonal Antibody (7H6F3E1)
GREENER_CHOICE

Catalog No. PIMA530025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530025 Supplier Invitrogen™ Supplier No. MA530025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human AACS/ACSF1.

Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AACS
Applications ELISA
Classification Monoclonal
Clone 7H6F3E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene AACS
Gene Accession No. Q86V21
Gene Alias 2210408B16Rik; aacs; AcAc-CoA ligase; acetoacetate-CoA ligase; acetoacetyl-CoA ligase; acetoacetyl-CoA synthetase; ACSF1; acyl-CoA synthetase family member 1; homolog of C. elegans supressor of ras 5 (sur-5); protein sur-5 homolog; si:dkey-215k6.2; SUR5; SUR-5; zgc:56105
Gene Symbols AACS
Host Species Mouse
Immunogen Recombinant Human AACS protein (Met1-Phe672).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.