Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AADAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542693 Supplier Invitrogen™ Supplier No. PA542693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AHLKTIQNLA TFVELLGLHH FMDSFKVVGS FDEVPPTSDE NVTVTETKFN Sequence homology: Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 91%; Rabbit: 100%; Rat: 90%.

Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens.Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens.Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AADAC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AADAC
Gene Accession No. P22760
Gene Alias 5033417E09Rik; Aada; AADAC; AI265437; Arylacetamide deacetylase; arylacetamide deacetylase (esterase); CES5A1; DAC
Gene Symbols AADAC
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human AADAC (aa 38-87).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.