Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AADACL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17931820UL
Description
AADACL1 Polyclonal specifically detects AADACL1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| AADACL1 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_848887 | |
| NCEH1 | |
| Synthetic peptide directed towards the middle region of human Aadacl1The immunogen for this antibody is Aadacl1. Peptide sequence LQALDFNTPSYQQSMNTPILPRHVMVRYWLDYFKGNYDFVEAMIVNNHTS. | |
| 20 μL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| AADACL1, EC 3.1.1.79, KIAA1363EC 3.1.1, NCEHArylacetamide deacetylase-like 1EC 3.1.1.-, neutral cholesterol ester hydrolase 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57552 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction