Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AADACL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7229739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB159620 Supplier Novus Biologicals Supplier No. NBP317795100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AADACL1 Polyclonal antibody specifically detects AADACL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen AADACL1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias AADACL1, EC 3.1.1.79, KIAA1363EC 3.1.1, NCEHArylacetamide deacetylase-like 1EC 3.1.1.-, neutral cholesterol ester hydrolase 1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: SDPWKLMLLDATFRGAQQVSNLIHYLGLSHHLLALNFIIVSFGKKSAWSSAQVKVTDTDFDGVEVRVFEGPPKPEEPLKRSVVYIHGGGWALASAKIRYYDELCTAMAEELNAVIVSIEYRLVPKVYFPEQIHDVVRATKYFLKP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57552
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.