Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AADACL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549736 Supplier Invitrogen™ Supplier No. PA549736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous level of total AADACL4 protein.

Arylacetamide deacetylase-like 4 is a protein in humans that is encoded by the AADACL4 gene. It belongs to the 'GDXG' lipolytic enzyme family. The subcellular location of this protein in the membrane, it is single-pass type II membrane protein. It has signal-anchor, transmembrane and transmembrane helix domain. Functions as Hydrolase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AADACL4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene AADACL4
Gene Accession No. Q5VUY2
Gene Alias Aadacl4; arylacetamide deacetylase like 4; arylacetamide deacetylase-like 4; Gm13177; OTTMUSG00000010747; predicted gene 13177; RGD1565761
Gene Symbols AADACL4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from 71-85 amino acids of human arylacetamide deacetylase-like 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343066
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.