Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AADAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532646
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532646 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532646 Supplier Invitrogen™ Supplier No. PA532646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Monkey (88%); Dog, Elephant (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AADAT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Aadat
Gene Accession No. Q8N5Z0
Gene Alias 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AadAT; Aadt; AI875679; alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; kynurenine aminotransferase II; kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2
Gene Symbols Aadat
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human AADAT.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51166
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.