Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AADAT Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142327
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
Specifications
| AADAT | |
| Polyclonal | |
| Unconjugated | |
| Aadat | |
| 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AadAT; Aadt; AI875679; alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; kynurenine aminotransferase II; kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51166 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8N5Z0 | |
| Aadat | |
| Peptide with sequence C-DRVIDFYSNQKD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction