Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554374 Supplier Invitrogen™ Supplier No. PA554374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFLQEANVTL IVIGQSSYHH IEPFCKLTGY SHEIYVDPER EIYKRLGMKR GEEIASSGQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AAED1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AAED1
Gene Accession No. Q7RTV5
Gene Alias 1110018J18Rik; Aaed1; AhpC/TSA antioxidant enzyme domain containing 1; ahpC/TSA antioxidant enzyme domain-containing protein 1; AI849003; C9orf21; Peroxiredoxin-like 2C; Prxl2c; Thioredoxin-like protein AAED1; UPF0308 protein C9orf21; UPF0308 protein C9orf21 homolog
Gene Symbols AAED1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AAED1. Recombinant protein control fragment (Product #RP-93022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 195827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.