Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAGAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546781 Supplier Invitrogen™ Supplier No. PA546781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLNSLTGTNH SIGSADPCHP EQPHLPAADS TESLSDHRGG ASNTTDAQVD.

The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAGAB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AAGAB
Gene Accession No. Q6PD74
Gene Alias 2310007F21Rik; AAGAB; alpha- and gamma-adaptin binding protein; alpha- and gamma-adaptin-binding protein p34; KPPP1; p34; PPKP1; PPKP1A
Gene Symbols AAGAB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human AAGAB (aa 186-235).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79719
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.