Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAGAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559090 Supplier Invitrogen™ Supplier No. PA559090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGASNTTDAQ VDSIVDPMLD LDIQELASLT TGGGDVENFE RLFSKLKEMK DKAATLPHEQ RKVHAEKVAK AFWMAIG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAGAB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AAGAB
Gene Accession No. Q6PD74
Gene Alias 2310007F21Rik; AAGAB; alpha- and gamma-adaptin binding protein; alpha- and gamma-adaptin-binding protein p34; KPPP1; p34; PPKP1; PPKP1A
Gene Symbols AAGAB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AAGAB. Recombinant protein control fragment (Product #RP-98027).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79719
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.