Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | AAK1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17957420
![]() |
Novus Biologicals
NBP17957420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179574
![]() |
Novus Biologicals
NBP179574 |
100 μL |
Each for $487.50
|
|
|||||
Description
AAK1 Polyclonal specifically detects AAK1 in Mouse samples. It is validated for Western Blot.Specifications
AAK1 | |
Polyclonal | |
Rabbit | |
NP_001035195 | |
22848 | |
The immunogen for this antibody is Aak1. Peptide sequence APVRQQPKVQTTPPPTIQGQKVGSLTPPSSPKTQRAGHRRILSDVTHSAV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570 | |
AAK1 | |
IgG | |
103 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title