Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582606 Supplier Invitrogen™ Supplier No. PA582606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLNKDFAKLG EGKHPEKLGG SAESLIPGFQ STQGDAFATT SFSAGTAEKR KGGQTVDSGL PLLSVSDPFI PLQVPDAPEK LIEGLKSPDT SLLLPDLLPM TDPFGSTSDA VIGKVIISVS SVMHDMCA Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 86%.

AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis. AAK1 is a member of the SNF1 subfamily of Ser/Thr protein kinases. The protein interacts with and phosphorylates a subunit of the AP-2 complex, which promotes binding of AP-2 to sorting signals found in membrane-bound receptors and subsequent receptor endocytosis. Its kinase activity is stimulated by clathrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AAK1
Gene Accession No. Q2M2I8
Gene Alias 5530400K14Rik; 9630042K20; Aak1; Adaptor-associated kinase 1; AP2 associated kinase 1; AP2-associated protein kinase 1; AU067724; AU067726; BC028270; C79663; D6Ertd245e; DKFZp686F03202; DKFZp686K16132; FLJ23712; FLJ25931; FLJ31060; FLJ42882; FLJ45252; KIAA1048; MGC138170; MGC164568; MGC164570; mKIAA1048; R75501; RGD1563580
Gene Symbols AAK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AAK1. Recombinant protein control fragment (Product #RP-91417).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.