Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAMDC Antibody (OTI1C4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271840FR
Description
AAMDC Monoclonal specifically detects AAMDC in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
AAMDC | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
AAMDC | |
Full length recombinant protein of human C11orf67 (NP_078960) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1C4 | |
Western Blot | |
adipogenesis associated, Mth938 domain containing, C11orf67, chromosome 11 open reading frame 67, CK067, FLJ21035, hypothetical protein LOC28971, MGC3367, PTD015 | |
Mouse | |
Affinity Purified | |
metabolism | |
28971.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction